Assay Details
Gene Symbol: CDC42-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF086337.1 | - | 1 | 255 | 184 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | W75995.1 | - | 256 | Chr. - - [-] on Build | 184 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 339 | LINC00339 |
HSPC157, NCRNA00339 |
-28.0kb | Sense | Chr. 1 - 22025191 - 22031224 on NCBI Build GRCh38 |
Downstream | Wnt family member 4 | WNT4 |
SERKAL, WNT-4 |
57.6kb | Anti-sense | Chr. 1 - 22117305 - 22143981 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
