Assay Details
Gene Symbol: LOC105372793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC042463.1 | - | 1 | 2456 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | potassium voltage-gated channel subfamily E regulatory subunit 1 | KCNE1 |
ISK, JLNS, JLNS2, LQT2/5, LQT5, MinK |
Anti-sense | Chr. 21 - 34446688 - 34512275 on NCBI Build GRCh38 | |
Downstream | regulator of calcineurin 1 | RCAN1 |
ADAPT78, CSP1, DSC1, DSCR1, MCIP1, RCN1 |
91.1kb | Anti-sense | Chr. 21 - 34516442 - 34615142 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
