Assay Details
Gene Symbol: VANGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172411.1 | NP_001165882.1 | 6 - 7 | 1343 | 76 | |
NM_001172412.1 | NP_001165883.1 | 6 - 7 | 1293 | 76 | ||
NM_138959.2 | NP_620409.1 | 6 - 7 | 1349 | 76 | ||
GenBank mRNA | AB057596.1 | - | 6 - 7 | 1293 | 76 | |
AB075805.1 | - | 5 - 6 | 1090 | 76 | ||
AF481859.1 | - | 6 - 7 | 1305 | 76 | ||
BC032773.1 | - | 4 - 5 | 1017 | 76 | ||
BC065272.1 | - | 6 - 7 | 1326 | 6165098 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
