Assay Details
Gene Symbol: CENPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110215.2 | NP_001103685.1 | 1 - 2 | 83 | 87 | |
GenBank mRNA | EU035297.1 | - | 1 - 2 | 83 | 87 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000836.hg.1
Additional Information:
This assay is in the 5'UTR region of the EU035297.1,NM_001110215.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR22014109.hg.1 | ENST00000472374.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22014109.hg.1 | NM_001110215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22014109.hg.1 | TC22000836.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000215980.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000215980.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000215980.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000215980.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000215980.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000215980.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000215980.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000215980.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000396437.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000396437.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000396437.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000396437.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000396437.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000396437.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000402338.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000402338.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000402338.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000402338.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000402338.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000402338.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000402338.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000402420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000402420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000402420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000402420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000402420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000402420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000402420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000404067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000404067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000404067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000404067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000404067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000404067.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000407253.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000407253.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000407253.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000407253.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000407253.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000407253.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000407253.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000460824.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000460824.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000460824.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000460824.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000472374.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000472374.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000472374.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | ENST00000472374.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001002876.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001002876.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001002876.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001002876.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001002876.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001002876.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001002876.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001110215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001110215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001110215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001110215.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304370.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304370.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304370.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304370.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304370.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304370.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304370.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304370.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304370.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304373.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304373.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304373.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304373.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304373.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304373.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304373.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_001304373.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_024053.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_024053.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_024053.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_024053.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_024053.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_024053.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_024053.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008847.hg.1 | NM_024053.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200164102.hg.1 | TC2200008847.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200164102.hg.1 | ENST00000472374.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200164102.hg.1 | NM_001110215.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200164102.hg.1 | ENST00000472374.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200164102.hg.1 | NM_001110215.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200164103.hg.1 | TC2200008847.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200164103.hg.1 | ENST00000472374.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200164103.hg.1 | NM_001110215.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200164103.hg.1 | ENST00000472374.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200164103.hg.1 | NM_001110215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000215980.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000215980.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000215980.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000215980.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000215980.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000215980.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000215980.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000215980.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000396437.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000396437.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000396437.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000396437.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000396437.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000396437.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000402338.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000402338.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000402338.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000402338.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000402338.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000402338.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000402338.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000402420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000402420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000402420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000402420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000402420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000402420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000402420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000404067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000404067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000404067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000404067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000404067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000404067.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000407253.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000407253.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000407253.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000407253.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000407253.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000407253.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000407253.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000460824.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000460824.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000460824.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000460824.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000472374.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000472374.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000472374.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | ENST00000472374.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001002876.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001002876.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001002876.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001002876.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001002876.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001002876.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001002876.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001110215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001110215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001110215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001110215.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304370.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304370.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304370.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304370.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304370.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304370.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304370.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304370.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304370.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304373.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304373.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304373.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304373.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304373.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304373.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304373.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_001304373.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_024053.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_024053.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_024053.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_024053.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_024053.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_024053.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_024053.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008847.hg.1 | NM_024053.4 | Refseq |
