Assay Details
Gene Symbol: SENP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172109.1 | NP_001165580.1 | 2 - 3 | 443 | 187 | |
GenBank mRNA | DB048641.1 | - | 2 - 3 | 443 | 187 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001172109.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
