Assay Details
Gene Symbol: SENP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172111.1 | NP_001165582.1 | 2 - 3 | 302 | 200 | |
GenBank mRNA | CN367895.1 | - | 2 - 3 | 269 | 200 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15000654.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001172111.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR15005613.hg.1 | ENST00000340912.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15005613.hg.1 | ENST00000542035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15005613.hg.1 | ENST00000544171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15005613.hg.1 | ENST00000544411.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15005613.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15005613.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15005613.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15005613.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15005613.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15005613.hg.1 | TC15000654.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR15005612.hg.1 | ENST00000544171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15005612.hg.1 | ENST00000544411.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15005612.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15005612.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15005612.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15005612.hg.1 | TC15000654.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | TC1500007777.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155206.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155205.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155205.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155205.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155205.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155205.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155205.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155205.hg.1 | TC1500007777.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500155205.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155205.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155205.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155205.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155205.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155205.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
