Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000468375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000468375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000468375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000468375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000488588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000488588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000488588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000488588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000488588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000494455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000494455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000494455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000494455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000494455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
ENST00000494455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
NM_014477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
NM_014477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
NM_014477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
NM_014477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
NM_014477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
NM_014477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
NM_014477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009220.hg.1 |
NM_014477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011992.hg.1 |
TC20000879.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011992.hg.1 |
ENST00000372726.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011992.hg.1 |
ENST00000494455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011992.hg.1 |
ENST00000537995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011992.hg.1 |
NM_014477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164503.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164503.hg.1 |
ENST00000488588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164503.hg.1 |
ENST00000494455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164503.hg.1 |
NM_014477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164503.hg.1 |
TC2000009220.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164503.hg.1 |
NM_014477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164503.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164503.hg.1 |
ENST00000488588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164503.hg.1 |
ENST00000494455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000372726.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000468375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000468375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000468375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000468375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000488588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000488588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000488588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000488588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000488588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000494455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000494455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000494455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000494455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000494455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
ENST00000494455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
NM_014477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
NM_014477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
NM_014477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
NM_014477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
NM_014477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
NM_014477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
NM_014477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009220.hg.1 |
NM_014477.2 |
Refseq |