Assay Details
Gene Symbol: IL22RA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052962.2 | NP_443194.1 | 6 - 7 | 945 | 184 | |
NM_181309.1 | NP_851826.1 | 5 - 6 | 849 | 184 | ||
GenBank mRNA | AJ297262.1 | - | 4 - 5 | 569 | 184 | |
AJ313161.1 | - | 5 - 6 | 851 | 184 | ||
AJ313162.1 | - | 6 - 7 | 947 | 184 | ||
AY040566.1 | - | 5 - 6 | 822 | 184 | ||
AY040567.1 | - | 6 - 7 | 918 | 184 | ||
AY044429.1 | - | 5 - 6 | 784 | 184 | ||
AY358111.1 | - | 5 - 6 | 847 | 184 | ||
AY358737.1 | - | 6 - 7 | 881 | 184 | ||
BC125167.1 | - | 4 - 5 | 611 | 40121609 | 184 | |
BC125168.1 | - | 4 - 5 | 611 | 40121612 | 184 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06002151.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06027118.hg.1 | TC06002151.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06027118.hg.1 | ENST00000296980.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027118.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027118.hg.1 | ENST00000349184.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027118.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027118.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027118.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027117.hg.1 | TC06002151.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06027117.hg.1 | ENST00000296980.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027117.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027117.hg.1 | ENST00000349184.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027117.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027117.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027117.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013299.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027120.hg.1 | TC06002151.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06027120.hg.1 | ENST00000296980.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027120.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027120.hg.1 | ENST00000349184.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027120.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027120.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027120.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192034.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192034.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192034.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192034.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192034.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192034.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192034.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192034.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192034.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192034.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192034.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192034.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192034.hg.1 | TC0600013299.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600192036.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192036.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192036.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192036.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192036.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192036.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192036.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192036.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192036.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192036.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192036.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192036.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192036.hg.1 | TC0600013299.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013299.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192035.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192035.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192035.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192035.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192035.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192035.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192035.hg.1 | ENST00000296980.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192035.hg.1 | ENST00000339602.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192035.hg.1 | ENST00000349184.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192035.hg.1 | NM_052962.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192035.hg.1 | NM_181309.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192035.hg.1 | NM_181310.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192035.hg.1 | TC0600013299.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 131735 | - | - |
