Assay Details
Gene Symbol: MIR455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030255.1 | 1 | 48 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | kinesin family member 12 | KIF12 | -109.4kb | Anti-sense | Chr. 9 - 114091623 - 114100099 on NCBI Build GRCh38 | |
Downstream | orosomucoid 1 | ORM1 |
AGP-A, AGP1, HEL-S-153w, ORM |
113.5kb | Sense | Chr. 9 - 114323023 - 114326479 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900008506.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09000581.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910836 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27254871 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910829 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27254844 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910817 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910793 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910788 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910804 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910835 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16255848 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910802 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910820 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910831 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910813 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910824 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27254861 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910800 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910783 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27254836 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910791 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910799 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910818 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910786 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27254829 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27254828 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910787 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910816 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910789 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910784 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910823 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910832 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27254863 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910797 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910795 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910834 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27254869 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27254876 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16255849 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910809 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910811 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910812 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910806 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910799 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910793 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910788 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910816 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910818 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910802 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910789 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27254844 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27254863 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910783 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910787 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27254876 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910811 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910832 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910817 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910800 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910813 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27254869 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910824 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910836 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910823 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910806 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910809 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27254871 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910829 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16255849 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910820 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27254828 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27254836 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910835 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910831 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910791 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910797 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910784 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910795 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910812 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27254861 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910786 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910834 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16255848 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27254829 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06910804 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.1 ST Array | 8157444 | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR09006621.hg.1 | ENST00000384993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09006621.hg.1 | NR_030255.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09006621.hg.1 | TC09000581.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0900008506.hg.1 | ENST00000384993.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008506.hg.1 | NR_030255.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008506.hg.1 | ENST00000384993.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008506.hg.1 | NR_030255.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900156949.hg.1 | ENST00000384993.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900156949.hg.1 | NR_030255.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900156949.hg.1 | TC0900008506.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900156949.hg.1 | ENST00000384993.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900156949.hg.1 | NR_030255.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3309919 | NR_030255.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3309919 | NR_030255.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309919 | NR_030255.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309919 | NR_030255.1 | human_transcriptome |
