Assay Details
Gene Symbol: MIR587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030314.1 | 1 | 44 | 76 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC100422737 | LOC100422737 | Anti-sense | Chr. 6 - 106717452 - 106787425 on NCBI Build GRCh38 | ||
Downstream | CD24 molecule | CD24 | CD24A | 185.6kb | Anti-sense | Chr. 6 - 106969831 - 106976855 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600008997.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
