Assay Details
Gene Symbol: MIR4294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_036182.1 | 1 | 33 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | WDFY family member 4 | WDFY4 | C10orf64 | -2.6kb | Anti-sense | Chr. 10 - 48684815 - 48982956 on NCBI Build GRCh38 |
Downstream | V-set and transmembrane domain containing 4 | VSTM4 | C10orf72 | 28.7kb | Sense | Chr. 10 - 49014245 - 49129118 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000010588.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001270.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
