Assay Details
Gene Symbol: MIR2355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_036227.1 | 1 | 48 | 75 |
Gene Symbol: KLF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017005163.1 | XP_016860652.1 | 3 | 5319 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | carboxypeptidase O | CPO | -138.9kb | Anti-sense | Chr. 2 - 206932853 - 206971064 on NCBI Build GRCh38 | |
Downstream | microRNA 7845 | MIR7845 | hsa-mir-7845 | 56.3kb | Anti-sense | Chr. 2 - 207166400 - 207166498 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200015539.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02002706.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
