Assay Details
Gene Symbol: MIR4290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_036253.1 | 1 | 41 | 83 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101927847 | LOC101927847 | -36.0kb | Anti-sense | Chr. 9 - 89969416 - 89987486 on NCBI Build GRCh38 | |
Downstream | long intergenic non-protein coding RNA 1508 | LINC01508 | 277.4kb | Sense | Chr. 9 - 90300896 - 90433489 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900010724.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09001316.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
