Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015585.hg.1 |
ENST00000374247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015585.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015585.hg.1 |
TC10001252.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015584.hg.1 |
ENST00000224605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015584.hg.1 |
ENST00000374247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015584.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015584.hg.1 |
TC10001252.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015582.hg.1 |
ENST00000224605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015582.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015582.hg.1 |
TC10001252.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000151329.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000151329.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000151329.hg.1 |
TC1000007543.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000151329.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000151329.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000151328.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000151328.hg.1 |
TC1000007543.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000151328.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000151331.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000151331.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000151331.hg.1 |
TC1000007543.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000151331.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000151331.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
ENST00000580279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007543.hg.1 |
NM_004962.4 |
Refseq |