Assay Details
Gene Symbol: GDF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004962.3 | NP_004953.1 | 2 - 3 | 1670 | 76 | |
GenBank mRNA | AK315322.1 | - | 2 - 3 | 1523 | 76 | |
BC028237.1 | - | 2 - 3 | 1100 | 5245100 | 76 | |
D49492.1 | - | 2 - 3 | 1508 | 76 | ||
L42113.1 | - | 1 - 2 | 165 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
