Assay Details
Gene Symbol: CD8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001768.6 | NP_001759.3 | 1 | 418 | 94 | |
NM_171827.3 | NP_741969.1 | 1 | 418 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001768.6, NM_171827.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
