Assay Details
Gene Symbol: MIR888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030592.1 | 1 | 44 | 76 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 890 | MIR890 |
MIRN890, hsa-mir-890 |
-433.0 | Sense | Chr. X - 145994275 - 145994351 on NCBI Build GRCh38 |
Downstream | microRNA 892a | MIR892A | MIRN892A | 1.8kb | Sense | Chr. X - 145996669 - 145996743 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00011030.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X001452.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
