Assay Details
Gene Symbol: MIR887 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030616.1 | 1 | 42 | 53 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101929472 | CTD-2350J17.1 | -320.2kb | Anti-sense | Chr. 5 - 15602189 - 15615002 on NCBI Build GRCh38 | |
Downstream | membrane associated ring-CH-type finger 11 | MARCH11 | MARCH-XI | 132.1kb | Anti-sense | Chr. 5 - 16067365 - 16179788 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
