Assay Details
Gene Symbol: MIR2114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031748.1 | 1 | 47 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 894 | LINC00894 | -211.2kb | Sense | Chr. X - 149938548 - 150016787 on NCBI Build GRCh38 | |
Downstream | mastermind like domain containing 1 | MAMLD1 |
CG1, CXorf6, F18, HYSP2 |
133.5kb | Sense | Chr. X - 150361422 - 150514178 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00008709.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000720.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021837 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757824 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021843 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757826 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021867 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021878 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757849 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757863 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757843 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021852 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021860 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021839 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021874 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757834 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021857 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17411273 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757840 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757851 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021851 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757821 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021891 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17411268 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021838 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757867 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021886 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021873 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021888 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757801 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17411279 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17411270 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757845 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021856 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757820 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757815 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021849 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17411278 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021893 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021844 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757833 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757814 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021889 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021845 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757852 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021900 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021834 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021896 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17411275 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021898 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021876 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021836 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021866 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021864 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021869 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757808 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757843 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021878 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17411273 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021873 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021849 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757852 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021898 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021893 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757801 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021876 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17411270 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021864 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021839 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021856 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021837 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757849 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17411279 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021857 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021860 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021843 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021889 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17411275 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757814 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021891 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021867 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17411268 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021874 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757863 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757867 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021844 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021852 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021869 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757851 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021845 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757833 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757840 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021866 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021838 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021886 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021836 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021834 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021896 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757808 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757820 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757821 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757826 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757845 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021888 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021900 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757834 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12021851 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757815 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17411278 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32757824 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X009239.hg.1 | TC0X000720.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009239.hg.1 | ENST00000516645.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009239.hg.1 | NR_031748.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008709.hg.1 | ENST00000516645.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008709.hg.1 | NR_031748.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008709.hg.1 | ENST00000516645.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008709.hg.1 | NR_031748.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159123.hg.1 | ENST00000516645.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159123.hg.1 | NR_031748.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159123.hg.1 | ENST00000516645.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159123.hg.1 | NR_031748.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159123.hg.1 | TC0X00008709.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3310770 | NR_031748.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3310770 | NR_031748.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310770 | NR_031748.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310770 | NR_031748.1 | human_transcriptome |
