Assay Details
Gene Symbol: MIR2276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031753.1 | 1 | 48 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
ankyrin repeat domain 20 family member A19, pseudogene |
ANKRD20A19P | -213.1kb | Anti-sense | Chr. 13 - 23907284 - 23949315 on NCBI Build GRCh38 | |
Downstream | SPATA13 antisense RNA 1 | SPATA13-AS1 | 90.2kb | Anti-sense | Chr. 13 - 24252749 - 24254439 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300006603.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000052.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
