Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07014496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07014496.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07014496.hg.1 |
TC07000936.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07014498.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07014498.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07014498.hg.1 |
TC07000936.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07014500.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07014500.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07014500.hg.1 |
TC07000936.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07014501.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07014501.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07014501.hg.1 |
TC07000936.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009496.hg.1 |
ENST00000421648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009496.hg.1 |
ENST00000421648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009496.hg.1 |
ENST00000421648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009496.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009496.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009496.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009496.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009496.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700167522.hg.1 |
ENST00000421648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167522.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167522.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167522.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700167522.hg.1 |
TC0700009496.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700167522.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700167522.hg.1 |
ENST00000421648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167522.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167522.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009496.hg.1 |
ENST00000421648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009496.hg.1 |
ENST00000421648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009496.hg.1 |
ENST00000421648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009496.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009496.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009496.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009496.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009496.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009496.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700167519.hg.1 |
ENST00000421648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167519.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167519.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167519.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700167519.hg.1 |
TC0700009496.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700167519.hg.1 |
ENST00000421648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167519.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167519.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167519.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700167527.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167527.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167527.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700167527.hg.1 |
TC0700009496.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700167527.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167527.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167527.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700167526.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167526.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167526.hg.1 |
NR_033897.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700167526.hg.1 |
TC0700009496.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700167526.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167526.hg.1 |
ENST00000429289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167526.hg.1 |
NR_033897.1 |
Refseq |