Assay Details
Gene Symbol: LOC100129316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033912.1 | 2 - 3 | 1773 | 87 | ||
GenBank mRNA | AK128703.1 | - | 2 - 3 | 1773 | 87 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | spleen associated tyrosine kinase | SYK | p72-Syk | -168.2kb | Anti-sense | Chr. 9 - 90801680 - 90898560 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 484 | LINC00484 | C9orf73 | 49.0kb | Anti-sense | Chr. 9 - 91119062 - 91182762 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09001321.hg.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
