Assay Details
Gene Symbol: LOC100129316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033912.1 | 3 - 4 | 1933 | 91 | ||
GenBank mRNA | AK128703.1 | - | 3 - 4 | 1933 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | spleen associated tyrosine kinase | SYK | p72-Syk | -164.7kb | Anti-sense | Chr. 9 - 90801680 - 90898560 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 484 | LINC00484 | C9orf73 | 51.7kb | Anti-sense | Chr. 9 - 91119062 - 91182762 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09001321.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813784 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813860 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154164 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16239144 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16239138 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154175 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813842 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813851 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154168 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154241 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813798 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154177 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813855 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16239135 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813866 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154191 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154227 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813803 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16239148 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154276 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813854 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154180 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813825 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154162 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16239161 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813767 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154200 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154188 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16239143 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154266 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154253 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813840 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813801 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813845 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154202 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813820 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813821 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813837 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813785 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813853 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813780 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813852 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813864 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813802 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813806 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154267 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154243 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813849 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813769 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154207 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813779 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813836 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813818 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813781 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813765 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154271 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154239 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813775 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154269 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813828 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813807 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16239149 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16239158 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813809 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16239159 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813859 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154236 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16239150 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154258 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813808 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813771 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813839 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813792 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16239137 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154211 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154159 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813816 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813846 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813778 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16239162 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813844 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813817 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154218 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813796 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154270 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813824 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813787 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154176 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154169 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154282 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813827 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813847 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154273 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813789 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154186 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154285 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154203 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813788 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154284 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154181 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813794 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154232 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813872 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154166 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27154216 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813835 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813776 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06813868 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 208907 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 150271 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 335110 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 104954 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 207517 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3217278 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 6670 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3217278 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3217278 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 71096 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P32471444 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02131095 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18691267 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P32471444 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09016458.hg.1 | NR_033912.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09016458.hg.1 | TC09001321.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0900173157.hg.1 | NR_033912.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900173157.hg.1 | TC0900010755.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900173157.hg.1 | NR_033912.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037684 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037684 | - | - |
