Assay Details
Gene Symbol: MIR3646 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037419.1 | 1 | 36 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | HNF4A antisense RNA 1 | HNF4A-AS1 | uc002xlx | -12.4kb | Anti-sense | Chr. 20 - 44377743 - 44395706 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 1430 | LINC01430 | 40.6kb | Anti-sense | Chr. 20 - 44448777 - 44450557 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000007453.hg.1
This assay targets the same location as HTA transcript cluster(s) TC20000331.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
