Assay Details
Gene Symbol: LINC01268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038863.2 | 2 - 3 | 427 | 97 | ||
GenBank mRNA | BC037331.1 | - | 2 - 3 | 432 | 97 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | myristoylated alanine rich protein kinase C substrate | MARCKS |
80K-L, MACS, PKCSL, PRKCSL |
-4.5kb | Anti-sense | Chr. 6 - 113857335 - 113863475 on NCBI Build GRCh38 |
Downstream | uncharacterized FLJ34503 | FLJ34503 | 33.7kb | Anti-sense | Chr. 6 - 113904387 - 113921642 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06002034.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196870 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196842 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04997057 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04997041 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04997027 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04997029 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04997038 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196877 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196837 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196836 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196866 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196855 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196876 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196875 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196872 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04997014 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196882 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196835 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04997008 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196847 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04997016 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196878 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196848 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196861 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196874 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04997055 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196865 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04997017 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04997031 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196864 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196869 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196849 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196841 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196857 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196846 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196863 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196854 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04997030 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196844 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196856 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196832 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196858 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25196831 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04997037 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 759030 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3867584 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 40529 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3867584 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3867584 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P37757981 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P37757981 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P373703 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P37757981 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1557359_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR06025452.hg.1 | TC06002034.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06025452.hg.1 | ENST00000434296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06025452.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06025450.hg.1 | TC06002034.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06025450.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06025453.hg.1 | TC06002034.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06025453.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06025451.hg.1 | TC06002034.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06025451.hg.1 | ENST00000434296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06025451.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012925.hg.1 | ENST00000434296.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012925.hg.1 | ENST00000434296.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012925.hg.1 | ENST00000434296.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012925.hg.1 | ENST00000434296.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012925.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012925.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012925.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012925.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600189278.hg.1 | ENST00000434296.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600189278.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600189278.hg.1 | ENST00000434296.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600189278.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600189278.hg.1 | TC0600012925.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600189280.hg.1 | ENST00000434296.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600189280.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600189280.hg.1 | ENST00000434296.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600189280.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600189280.hg.1 | TC0600012925.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600012925.hg.1 | ENST00000434296.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012925.hg.1 | ENST00000434296.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012925.hg.1 | ENST00000434296.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012925.hg.1 | ENST00000434296.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012925.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012925.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012925.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012925.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600189276.hg.1 | ENST00000434296.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600189276.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600189276.hg.1 | ENST00000434296.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600189276.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600189276.hg.1 | TC0600012925.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600189277.hg.1 | ENST00000434296.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600189277.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600189277.hg.1 | ENST00000434296.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600189277.hg.1 | NR_038863.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600189277.hg.1 | TC0600012925.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 1557359_at | - | - |
Human Genome U133 Plus 2.0 Array | 1557359_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557359_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557359_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557359_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557359_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557359_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557359_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557359_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557359_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557359_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557359_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557359_at3 | - | - |
Human Genome U95 Set | 71130_at | - | - |
Human Genome U95 Set | 71130_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030412 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030413 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030413 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030412 | - | - |
