Assay Details
Gene Symbol: MIR4418 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_039613.1 | 1 | 33 | 61 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Wnt family member 4 | WNT4 |
SERKAL, WNT-4 |
-122.3kb | Anti-sense | Chr. 1 - 22117305 - 22143981 on NCBI Build GRCh38 |
Downstream | zinc finger and BTB domain containing 40 | ZBTB40 | ZNF923 | 162.5kb | Sense | Chr. 1 - 22428838 - 22531157 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100007276.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
