Assay Details
Gene Symbol: MIR4757 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_039914.1 | 1 | 40 | 60 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1376 | LINC01376 | -312.4kb | Anti-sense | Chr. 2 - 18986451 - 19026971 on NCBI Build GRCh38 | |
Downstream | odd-skipped related transciption factor 1 | OSR1 | ODD | 3.0kb | Anti-sense | Chr. 2 - 19351485 - 19358611 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200006853.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02000109.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
