Assay Details
Gene Symbol: A2MP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040112.1 | 8 - 9 | 1034 | 89 | ||
GenBank mRNA | DB301195.1 | - | 1 - 2 | 89 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
PZP, alpha-2-macroglobulin like |
PZP | CPAMD6 | -20.2kb | Sense | Chr. 12 - 9140730 - 9208370 on NCBI Build GRCh38 |
Downstream | microRNA 1244-3 | MIR1244-3 | 10.1kb | Sense | Chr. 12 - 9239467 - 9239551 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001189.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
