Assay Details
Gene Symbol: HIST1H2APS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045125.1 | 1 | 320 | 57 | ||
GenBank mRNA | DB441866.1 | - | 1 | 320 | 57 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
histone cluster 1, H2ba |
HIST1H2BA |
H2BFU, STBP, TH2B, TSH2B, TSH2B.1, bA317E16.3 |
-5.1kb | Sense | Chr. 6 - 25726909 - 25727345 on NCBI Build GRCh38 |
Downstream | solute carrier family 17 member 4 | SLC17A4 | KAIA2138 | 20.5kb | Sense | Chr. 6 - 25754673 - 25781175 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000159.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
