Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT HG-U133+ PM Array Plate |
228675_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025313.hg.1 |
ENST00000478808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025313.hg.1 |
NR_038996.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025313.hg.1 |
TC12001946.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025314.hg.1 |
ENST00000478808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025314.hg.1 |
NR_038996.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025314.hg.1 |
TC12001946.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025312.hg.1 |
NR_038996.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025312.hg.1 |
TC12001946.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200191968.hg.1 |
ENST00000478808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191968.hg.1 |
NR_038996.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191968.hg.1 |
TC1200011860.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200191968.hg.1 |
NR_038996.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191968.hg.1 |
ENST00000478808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191967.hg.1 |
ENST00000478808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191967.hg.1 |
NR_038996.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191967.hg.1 |
TC1200011860.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200191967.hg.1 |
ENST00000478808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191967.hg.1 |
NR_038996.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191966.hg.1 |
NR_038996.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191966.hg.1 |
TC1200011860.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200191966.hg.1 |
NR_038996.1 |
Refseq |
Human Genome U133 Plus 2.0 Array |
228675_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
228675_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
228675_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
228675_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
228675_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
228675_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
228675_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
228675_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
228675_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
228675_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
228675_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
228675_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
228675_at10 |
- |
- |
Human Genome U95 Set |
90437_at |
- |
- |
Human Genome U95 Set |
90437_PM_at |
- |
- |
Human Genome U95 Set |
68294_PM_at |
- |
- |
Human Genome U95 Set |
68294_at |
- |
- |
Human Genome U133 Set |
228675_PM_at |
- |
- |
Human Genome U133 Set |
228675_at |
- |
- |