Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22009393.hg.1 |
TC22000506.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22009393.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22009396.hg.1 |
TC22000506.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22009396.hg.1 |
ENST00000540078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22009396.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22009395.hg.1 |
TC22000506.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22009395.hg.1 |
ENST00000540078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22009395.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22009397.hg.1 |
TC22000506.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22009397.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200157336.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200157336.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200157336.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200157336.hg.1 |
TC2200008022.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200157336.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200157336.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200157336.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200157337.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200157337.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200157337.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200157337.hg.1 |
TC2200008022.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200157337.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200157337.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200157337.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200157338.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200157338.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200157338.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200157338.hg.1 |
TC2200008022.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200157338.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200157338.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200157338.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200157339.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200157339.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200157339.hg.1 |
TC2200008022.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200157339.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200157339.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |