Assay Details
Gene Symbol: LOC646736 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046102.1 | 3 - 4 | 443 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adaptor 2 | NYAP2 | KIAA1486 | -460.7kb | Sense | Chr. 2 - 225399710 - 225703653 on NCBI Build GRCh38 |
Downstream | microRNA 5702 | MIR5702 | 493.5kb | Anti-sense | Chr. 2 - 226658710 - 226658793 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001358.hg.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
