Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150505.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150505.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150505.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150505.hg.1 |
TC0300007170.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300150505.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150505.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150505.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150504.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150504.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150504.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150504.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150504.hg.1 |
TC0300007170.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300150504.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150504.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150504.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150504.hg.1 |
NR_046732.1 |
Refseq |