Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT HG-U133+ PM Array Plate |
1557652_PM_a_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007170.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150506.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150506.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150506.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150506.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150506.hg.1 |
TC0300007170.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300150506.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150506.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150506.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150506.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007170.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150507.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150507.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150507.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150507.hg.1 |
NR_046732.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150507.hg.1 |
TC0300007170.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300150507.hg.1 |
ENST00000418161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150507.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150507.hg.1 |
ENST00000446950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150507.hg.1 |
NR_046732.1 |
Refseq |
Human Genome U133 Plus 2.0 Array |
1557652_a_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1557652_PM_a_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557652_a_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557652_a_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557652_a_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557652_a_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557652_a_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557652_a_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557652_a_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557652_a_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557652_a_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557652_a_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557652_a_at6 |
- |
- |