Assay Details
Gene Symbol: AOAH-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046764.1 | 1 - 2 | 377 | 101 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | anillin actin binding protein | ANLN |
FSGS8, Scraps, scra |
-144.4kb | Anti-sense | Chr. 7 - 36389803 - 36453791 on NCBI Build GRCh38 |
Downstream | engulfment and cell motility 1 | ELMO1 |
CED-12, CED12, ELMO-1 |
252.8kb | Sense | Chr. 7 - 36852906 - 37449409 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
