Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008306.hg.1 |
ENST00000441845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008306.hg.1 |
ENST00000441845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008306.hg.1 |
ENST00000441845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008306.hg.1 |
ENST00000441845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008306.hg.1 |
NR_046832.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008306.hg.1 |
NR_046832.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008306.hg.1 |
NR_046832.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008306.hg.1 |
NR_046832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600158017.hg.1 |
ENST00000441845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600158017.hg.1 |
NR_046832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600158017.hg.1 |
ENST00000441845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600158017.hg.1 |
NR_046832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600158017.hg.1 |
TC0600008306.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600158016.hg.1 |
ENST00000441845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600158016.hg.1 |
NR_046832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600158016.hg.1 |
ENST00000441845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600158016.hg.1 |
NR_046832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600158016.hg.1 |
TC0600008306.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008306.hg.1 |
ENST00000441845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008306.hg.1 |
ENST00000441845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008306.hg.1 |
ENST00000441845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008306.hg.1 |
ENST00000441845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008306.hg.1 |
NR_046832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008306.hg.1 |
NR_046832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008306.hg.1 |
NR_046832.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008306.hg.1 |
NR_046832.1 |
Refseq |