Assay Details
Gene Symbol: MLIP-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046832.1 | 3 - 4 | 457 | 156 | ||
GenBank mRNA | CF890740.1 | - | 3 - 4 | 457 | 156 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101927189 | LOC101927189 | -4.9kb | Sense | Chr. 6 - 53929982 - 53997667 on NCBI Build GRCh38 | |
Downstream | muscular LMNA-interacting protein | MLIP | C6orf142 | 3.8kb | Sense | Chr. 6 - 54018916 - 54266325 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
