Assay Details
Gene Symbol: MIR5189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_049821.1 | 1 | 61 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
zinc finger protein, FOG family member 1 |
ZFPM1 |
FOG, FOG1, ZC2HC11A, ZNF408, ZNF89A |
Sense | Chr. 16 - 88451782 - 88535166 on NCBI Build GRCh38 | |
Downstream | ZC3H18 antisense RNA 1 (head to head) | ZC3H18-AS1 |
C16orf85, LINC00859 |
85.0kb | Anti-sense | Chr. 16 - 88553981 - 88567106 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600008824.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
