Assay Details
Gene Symbol: MAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002382.4 | NP_002373.3 | 4 - 5 | 500 | 66 | |
NM_145112.2 | NP_660087.1 | 3 - 4 | 473 | 66 | ||
NR_073137.1 | 3 - 4 | 419 | 66 | |||
GenBank mRNA | AK290130.1 | - | 3 - 4 | 404 | 66 | |
AK292189.1 | - | 4 - 5 | 410 | 66 | ||
BC003525.1 | - | 3 - 4 | 383 | 3607261 | 66 | |
BC004516.2 | - | 4 - 5 | 426 | 3937573 | 66 | |
BC025685.1 | - | 3 - 4 | 420 | 5205305 | 66 | |
BC027924.1 | - | 3 - 4 | 430 | 5309524 | 66 | |
M64240.1 | - | 4 - 5 | 322 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
