Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011900.hg.1 |
ENST00000374945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011900.hg.1 |
ENST00000374945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011900.hg.1 |
ENST00000374945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011900.hg.1 |
ENST00000374945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011900.hg.1 |
ENST00000374945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011900.hg.1 |
ENST00000374945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011900.hg.1 |
ENST00000374945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011900.hg.1 |
NR_073178.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011900.hg.1 |
NR_073178.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011900.hg.1 |
NR_073178.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011900.hg.1 |
NR_073178.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011900.hg.1 |
NR_073178.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011900.hg.1 |
NR_073178.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011900.hg.1 |
NR_073178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197022.hg.1 |
ENST00000374945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197022.hg.1 |
NR_073178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197022.hg.1 |
TC1700011900.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700197022.hg.1 |
NR_073178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197022.hg.1 |
ENST00000374945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197023.hg.1 |
ENST00000374945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197023.hg.1 |
NR_073178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197023.hg.1 |
TC1700011900.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700197023.hg.1 |
ENST00000374945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197023.hg.1 |
NR_073178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011900.hg.1 |
ENST00000374945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011900.hg.1 |
ENST00000374945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011900.hg.1 |
ENST00000374945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011900.hg.1 |
ENST00000374945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011900.hg.1 |
ENST00000374945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011900.hg.1 |
ENST00000374945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011900.hg.1 |
ENST00000374945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011900.hg.1 |
NR_073178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011900.hg.1 |
NR_073178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011900.hg.1 |
NR_073178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011900.hg.1 |
NR_073178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011900.hg.1 |
NR_073178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011900.hg.1 |
NR_073178.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011900.hg.1 |
NR_073178.1 |
Refseq |