Assay Details
Gene Symbol: GABRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270873.1 | NP_001257802.1 | 4 - 5 | 747 | 70 | |
NM_033223.4 | NP_150092.2 | 4 - 5 | 747 | 70 | ||
XM_011521430.2 | XP_011519732.1 | 4 - 5 | 741 | 70 | ||
XM_017022058.1 | XP_016877547.1 | 2 - 3 | 3232 | 70 | ||
XM_017022059.1 | XP_016877548.1 | 2 - 3 | 572 | 70 | ||
GenBank mRNA | S82769.1 | - | 4 - 5 | 525 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
