Assay Details
Gene Symbol: GABRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270873.1 | NP_001257802.1 | 5 - 6 | 834 | 86 | |
NM_033223.4 | NP_150092.2 | 5 - 6 | 834 | 86 | ||
XM_011521430.2 | XP_011519732.1 | 5 - 6 | 828 | 86 | ||
XM_017022058.1 | XP_016877547.1 | 3 - 4 | 3319 | 86 | ||
XM_017022059.1 | XP_016877548.1 | 3 - 4 | 659 | 86 | ||
GenBank mRNA | S82769.1 | - | 5 - 6 | 612 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000554786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000554786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000557596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000557596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145982.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145982.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145982.hg.1 | TC1500006645.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500145982.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145982.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000554786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000554786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000557596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000557596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145983.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145983.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145983.hg.1 | TC1500006645.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500145983.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145983.hg.1 | NM_001270873.1 | Refseq |
