Assay Details
Gene Symbol: PPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000310.3 | NP_000301.1 | 1 - 2 | 354 | 60 | |
XM_005271008.1 | XP_005271065.1 | 1 - 2 | 138 | 60 | ||
GenBank mRNA | AK075496.1 | - | 1 - 2 | 129 | 60 | |
AK310799.1 | - | 1 - 2 | 136 | 60 | ||
AK312287.1 | - | 1 - 2 | 129 | 60 | ||
BC008426.1 | - | 1 - 2 | 134 | 4249991 | 60 | |
BX441569.2 | - | 1 - 2 | 587 | 60 | ||
CR542053.1 | - | 1 - 2 | 122 | 60 | ||
DA504318.1 | - | 1 - 2 | 354 | 60 | ||
U44772.1 | - | 1 - 2 | 135 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
