Assay Details
Gene Symbol: EML4-ALK fusion | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB374363.1 | - | 1726 | 114 | ||
OM_EML4-ALK_E14A20_COSF477.0 | - | 72 | 88 | |||
OM_EML4-ALK_E14A20_COSF477_1.0 | - | 98 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
