Assay Details
Gene Symbol: LOC285696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK125192.1 | - | 2 - 3 | 1070 | 144 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | myosin X | MYO10 | -246.1kb | Sense | Chr. 5 - 16661907 - 16936276 on NCBI Build GRCh38 | |
Downstream | brain abundant membrane attached signal protein 1 | BASP1 |
CAP-23, CAP23, NAP-22, NAP22 |
681.0 | Anti-sense | Chr. 5 - 17216823 - 17276845 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
