Assay Details
Gene Symbol: FUOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098483.2 | NP_001091953.1 | 5 - 6 | 418 | 113 | |
NM_001301827.1 | NP_001288756.1 | 4 - 5 | 349 | 113 | ||
NM_001301828.1 | NP_001288757.1 | 5 - 6 | 625 | 113 | ||
XM_011539672.2 | XP_011537974.1 | 5 - 6 | 457 | 113 | ||
GenBank mRNA | AI333356.1 | - | 1 - 2 | 113 | ||
BM009909.1 | - | 4 - 5 | 281 | 113 | ||
BU849497.1 | - | 5 - 6 | 601 | 113 | ||
BX119427.1 | - | 5 - 6 | 344 | 113 | ||
HY163253.1 | - | 5 - 6 | 413 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
