Assay Details
Gene Symbol: TGFB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003239.3 | NP_003230.1 | 6 - 7 | 2214 | 64 | |
GenBank mRNA | AK122902.1 | - | 3 - 4 | 1466 | 64 | |
AK313564.1 | - | 6 - 7 | 1295 | 64 | ||
EF560714.1 | - | 6 - 7 | 1198 | 64 | ||
J03241.1 | - | 6 - 7 | 1347 | 64 | ||
X14149.1 | - | 6 - 7 | 1338 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
