Assay Details
Gene Symbol: CCL27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006664.3 | NP_006655.1 | 2 - 3 | 262 | 101 | |
GenBank mRNA | AB010445.1 | - | 2 - 3 | 262 | 101 | |
AF082393.1 | - | 2 - 3 | 203 | 101 | ||
AJ243542.1 | - | 2 - 3 | 227 | 101 | ||
BC148263.1 | - | 2 - 3 | 225 | 8860203 | 101 | |
BM719357.1 | - | 1 - 2 | 135 | 101 | ||
CA433929.1 | - | 1 - 2 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |
