Assay Details
Gene Symbol: ATP6AP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017971.2 | NP_001017971.1 | 7 - 8 | 1403 | 62 | |
XM_011543723.2 | XP_011542025.1 | 7 - 8 | 2050 | 62 | ||
XM_017010039.1 | XP_016865528.1 | 2 - 3 | 209 | 62 | ||
XM_017010040.1 | XP_016865529.1 | 2 - 3 | 114 | 62 | ||
XM_017010041.1 | XP_016865530.1 | 8 - 9 | 2128 | 62 | ||
XM_017010042.1 | XP_016865531.1 | 9 - 10 | 2034 | 62 | ||
XM_017010043.1 | XP_016865532.1 | 8 - 9 | 2117 | 62 | ||
XM_017010044.1 | XP_016865533.1 | 7 - 8 | 2076 | 62 | ||
XM_017010045.1 | XP_016865534.1 | 7 - 8 | 1628 | 62 | ||
XM_017010046.1 | XP_016865535.1 | 7 - 8 | 1709 | 62 | ||
XM_017010047.1 | XP_016865536.1 | 6 - 7 | 1998 | 62 | ||
GenBank mRNA | AK022625.1 | - | 7 - 8 | 1403 | 62 | |
BC093982.1 | - | 1 - 2 | 133 | 7939827 | 62 | |
BC112191.1 | - | 1 - 2 | 133 | 8327659 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
