Assay Details
Gene Symbol: CAST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104285.1 | 17 - 18 | 1653 | 67 | ||
GenBank mRNA | U31346.1 | - | 9 - 10 | 885 | 67 | |
U58996.2 | - | 17 - 18 | 1632 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101929710 | LOC101929710 | -141.6kb | Sense | Chr. 5 - 95962001 - 96631085 on NCBI Build GRCh38 | |
Downstream | endoplasmic reticulum aminopeptidase 2 | ERAP2 |
L-RAP, LRAP |
102.5kb | Sense | Chr. 5 - 96875940 - 96919703 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
